Cole-Parmer MP0415/MP(R)115/MP(R)315 Centrifuge Fixed Angle Rotor, 24 x Capillary and 12 x 2 mL
Representative image only.
Cole-Parmer Item # EW-17406-78
$388.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)12000
  • Min RCF x g13500
  • Rotor TypeFixed angle
  • Tube Sizes Accepted2 mL
  • Number Of Tubes Accepted12
  • DescriptionCentrifuge Fixed Angle Rotor, 24 x Capillary and 12 x 2 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.