This item is now discontinued. Please browse similar items
Cole-Parmer MP0415/MP(R)115/MP(R)315 Centrifuge Microtube Rotor, 4 x PCR Strip
Representative image only.
Cole-Parmer Item # EW-17406-31
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)15000
  • Min Speed (rpm)500
  • Max RCF x g22000
  • Rotor TypePCR strip
  • Tube Sizes Accepted0.1 to 0.2 mL
  • Number Of Tubes Accepted4
  • DescriptionCentrifuge Microtube Rotor, 4 x PCR Strip
More About this Item
Expand and change the capacity of your centrifuge with these rotors.