Cole-Parmer MP0415/MP(R)315 Centrifuge Microtube Rotor, 36 x 0.5 mL
Representative image only.
Cole-Parmer Item # EW-17406-28
$375.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)15000
  • Min RCF x g22000
  • Rotor TypeMicrotube
  • Tube Sizes Accepted0.5 mL
  • Number Of Tubes Accepted36
  • DescriptionCentrifuge Microtube Rotor, 36 x 0.5 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.