Cole-Parmer Multiconfigurable Pressure Transmitter w/o Display, 10-100 psi
Representative image only.
Cole-Parmer Item # EW-98073-80
Measure eight pressure ranges in a single unit
  • Choose models with LCD for on-site indication of pressure
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±1% full-scale; ±2% full-scale at lowest psi rating
  • Min Pressure (PSID)-100
  • Max Pressure (PSID)100
  • Wetted Materials17-4PH SS
  • Casealuminum
  • Process Connection1/8\" NPT(F)
  • Battery Type13 to 30 VDC/VAC, 3-wire
  • Electrical Connections2- and 3-wire
  • Output4 to 20 mA; 1 to 5, 0 to 5, 0 to 10 VDC
  • Enclosure RatingNEMA 4
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionMulticonfigurable Pressure Transmitter w/o Display, 10-100 psi
  • Warranty1 year
1 Year WarrantyISO9001:2000 Certified Supplier
Key Features
  • Choose models with LCD for on-site indication of pressure
  • 1 year warranty
More About this Item
These feature-rich, multiconfigurable wet-to-wet differential pressure transducers provides you with field-accessible pressure ranging, choice of output, and zeroing. Each unit has four unidirectional and four bidirectional switch-selectable ranges — read positive and negative gauge and differential pressure ranges, giving you eight pressure ranges in one transducer! All controls are contained in a NEMA 4 rated, cast aluminum housing that is easily accessible in the field.