Cole-Parmer n-Butyl Alcohol, ACS Grade; 1 L

Representative image only.
Cole-Parmer Item # EW-88405-94
Reagents and solutions to meet your lab and research needs
  • Assay - ≥99.8%
  • Color (APHA) - ≤5
  • Isobutanol - ≤0.1%
  • Water - ≤0.1%
  • Acidity - ≤0.005%

PRODUCT OPTIONS

$86.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationButanols
  • CAS Number71-36-3
  • UN NumberUN1120
  • Shelf Life (days)60
  • Descriptionn-Butyl Alcohol, ACS Grade, 1 L

Key Features

  • Assay - ≥99.8%
  • Color (APHA) - ≤5
  • Isobutanol - ≤0.1%
  • Water - ≤0.1%
  • Acidity - ≤0.005%
  • Aldehydes - None
  • Chlorides - None

More About this Item

Quality and consistency your lab can count on.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat