Cole-Parmer Oleic Acid Sodium Salt 1g

Representative image only.
Cole-Parmer Item # EW-89100-22
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • ChemistryAcids
  • Volume (mL)4000
  • Formula Weight282.46
  • CAS Number112-80-1
  • DescriptionOleic Acid Sodium Salt 1g
  • Warranty1 year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat