Cole-Parmer Orp probe, refillable/ptfe sj/epoxy/pt band/155x12mm; bnc
Representative image only.
Cole-Parmer Item # EW-05990-55
Unique probes for special applications
  • Combine quality with durability
  • Eposy-body housing resists impact
  • Compatible with most pH meters
  • 6 month warranty
PRODUCT OPTIONS
$245.00USD / Each
BUY MORE & SAVE
  • 1-3$245.00USD / Each
  • 4+$220.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionPTFE
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)80
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionOrp probe, refillable/ptfe sj/epoxy/pt band/155x12mm; bnc
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Combine quality with durability
  • Eposy-body housing resists impact
  • Compatible with most pH meters
  • 6 month warranty
More About this Item
These quality electrodes have a no maintenance sealed gel reference. All electrodes have a Ag/AgCl reference cell.
ORP special-purpose electrodes have a PTFE reference-junction that resists coating and clogging.