Cole-Parmer Paperless Recorder, 2-Channel; Isolated 12/24VDC Input and USB
Representative image only.
Cole-Parmer Item # EW-80818-02
No more paper to jam, no more pens to clog
  • Intuitive operation, flexible input configurations, and large memory capabilities
  • Highly reflective LCD with bright white LED backlight displays real-time data in digital or trend format
  • Two universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTDs, as well as frequency and pulse inputs
  • Four internal alarm set points, two alarm relay outputs, and one digital control input
  • Ethernet and USB communication options available
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Chart TypeDigital
  • Max Temperature (° F)4350
  • Min Temperature (° F)-150
  • Max Temperature (° C)2400
  • Min Temperature (° C)-101
  • Output TypeUSB
  • DescriptionPaperless Recorder, 2-Channel; Isolated 12/24VDC Input and USB
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Intuitive operation, flexible input configurations, and large memory capabilities
  • Highly reflective LCD with bright white LED backlight displays real-time data in digital or trend format
  • Two universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTDs, as well as frequency and pulse inputs
  • Four internal alarm set points, two alarm relay outputs, and one digital control input
  • Ethernet and USB communication options available
  • 1 year warranty
More About this Item
Compact size combined with extreme flexibility makes these two-channel paperless recorders a powerful, yet economical, digital data recording system. A maximum storage rate of 100 samples per second can be set for both channels, letting you capture short duration process signals anomalies. Use CompactFlash™ cards up to 2 GB for data storage over longer time periods.