Cole-Parmer PCR Plates, 384-well, unskirted
Representative image only.
Cole-Parmer Item # EW-67103-95
Designed to amplify nucleic acids during the PCR process
  • Precision molded from 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable.
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
PRODUCT OPTIONS
$248.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Capacity0.4 mL
  • DescriptionPCR Plates, 384-well, unskirted
Key Features
  • Precision molded from 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable.
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
More About this Item
Cole-Parmer PCR products are specifically designed for amplifying nucleic acids during the Polymerase Chain Reaction (PCR) process. Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.