PCRmax 96-well Polypropylene PCR Microplate, Full Skirt, Clear, Nonsterile; 50/Pk
Representative image only.
PCRmax Mfr # 321-60-052 – Item # EW-67103-66
Designed to amplify nucleic acids during the PCR process
  • Manufactured with 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
  • RNase-/DNase-free, nonpyrogenic, nonsterile
  • Single notch corner
  • Robotic friendly full skirt design
  • Can be sealed with sealing film
PRODUCT OPTIONS
$324.00USD / Case of 50
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.2
  • Well Volume (µL)200
  • Well ShapeRound
  • SterileNo
  • Plate TypeFull skirt
  • Plate ColorClear
  • MaterialPolypropylene
  • DescriptionPCR Plates, 96-well, Full Skirt
Key Features
  • Manufactured with 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
  • RNase-/DNase-free, nonpyrogenic, nonsterile
  • Single notch corner
  • Robotic friendly full skirt design
  • Can be sealed with sealing film
More About this Item
PCRmax® PCR products are specifically designed for amplifying nucleic acids during the Polymerase Chain Reaction (PCR) process. Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.