Cole-Parmer pH electrode, combination, rugged, glass body, interchangeble connection
Representative image only.
Cole-Parmer Item # EW-05658-05
Save space and money by using one cable with multiple electrodes or vice versa
  • Cable cap rotates independently and will not bunch up
  • 6 month warranty
PRODUCT OPTIONS
$185.00USD / Each
BUY MORE & SAVE
  • 1-3$185.00USD / Each
  • 4+$166.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsRugged lab and field
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorDepends upon cable purchased separately: BNC, US standard, or SN6 to 57
  • Diameter (mm)8
  • Length (mm)154
  • DescriptionpH electrode, combination, rugged, glass body, interchangeble connection
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Cable cap rotates independently and will not bunch up
  • 6 month warranty
More About this Item
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.