Cole-Parmer pH Electrode, double-junction, refillable, for use with flow cell 05662-40
Representative image only.
Cole-Parmer Item # EW-05662-45
Ideal for fermentation applications
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
PRODUCT OPTIONS
$296.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • ConnectorDepends upon cable purchased separately: BNC or Ingold K9
  • DescriptionpH Electrode, double-junction, refillable, for use with flow cell 05662-40
Key Features
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
More About this Item
Replacement electrodes for your flow-through cell. Select the electrode that is compatible with your model of cell, with sealed and refillable options available.