Cole-Parmer pH Electrodes for Field Use; single-junction 10 foot cable
Representative image only.
Cole-Parmer Item # EW-05992-04
Ideal for your most rugged applications
  • Ceramic reference-junction pH electrodes (clog resistant)
  • 1 year warranty
PRODUCT OPTIONS
$151.20USD / Each
$168.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Min pH (pH)14
  • Max Temperature (° C)100
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionpH Electrodes for Field Use; single-junction 10 foot cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Ceramic reference-junction pH electrodes (clog resistant)
  • 1 year warranty
More About this Item
These combination electrodes withstand the most demanding field conditions—the semi-domed tip protects the pH bulb and ensures a quick response when taking measurements in harsh and dirty environments. Easy-to-clean electrodes feature an impact-resistant epoxy body for further protection from the elements. The large, annular ceramic reference junction and sealed cross-linked polymer reference resist clogging.
Operating range is from 0 to 14 pH. Electrodes measure 110 mm L x 12 mm dia and withstand temperatures up to 212°F (100°C). All models have BNC connectors.