Cole-Parmer pH reference electrode, double junction, refillable, epoxy body, US Std
Representative image only.
Cole-Parmer Item # EW-05899-25
Applications include organics, metals, proteins, ISE, and aggressive chemicals measurement
  • Epoxy material
  • Half-cell
  • 1 year warranty
$254.00USD / Each
BUY MORE & SAVE
  • 1-3$254.00USD / Each
  • 4+$228.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsHalf-Cell. Organics, metals, proteins, ISE, and aggressive chemicals
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)80
  • ConnectorPin-tip (U.S. standard)
  • Diameter (mm)12
  • Length (mm)154
  • DescriptionpH reference electrode, double junction, refillable, epoxy body, US Std
  • Warranty1 year
1 Year Warranty
Key Features
  • Epoxy material
  • Half-cell
  • 1 year warranty
More About this Item
Order replacement filling solution below.