Cole-Parmer Phenolic Pegboard with Drip Trough, 53 Pegs, 30" x 30"

Representative image only.
Cole-Parmer Item # EW-33732-21
Ideal for drying beakers, test tubes, and other apparatus quickly

PRODUCT OPTIONS

$2,175.50USD / Each
Often Bought with...

SPECS & DESCRIPTION

Specifications & Description

  • Width (in)30
  • Height (in)30
  • Width (cm)76.2
  • Height (cm)76.2
  • DescriptionPhenolic Pegboard with Drip Trough, 53 Pegs, 30" x 30"

More About this Item

Phenolic resin pegboards are ideal for drying and storage of labware. Pegboards are 1” thick and include a stainless-steel drip trough to collect run-off.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat