Cole-Parmer Phenolphthalein, 1% (w/v) in Isopropyl Alcohol, 1 L

Representative image only.
Cole-Parmer Item # EW-88401-51
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

PRODUCT OPTIONS

$129.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC20H14O4
  • ClassChemical Indicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight318.32
  • PackagingPoly Bottle
  • DOT InformationIsopropanol Solution; Class 3, Packing Group II
  • CAS Number67-63-0, 77-09-8, 1310-73-2
  • UN NumberUN1219
  • Shelf Life (months)24
  • DescriptionPhenolphthalein, 1% (w/v) in Isopropyl Alcohol, 1 L

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat