Cole-Parmer Pinchcock Clamp, Heavy-Duty, Nickel-Plated Zinc, 0 to 0.43" Dia Tubing
Representative image only.
Cole-Parmer Item # EW-08126-02
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeTubing
  • Min Grip Range (in)0
  • Max Grip Range (in)0.43
  • Length (in)2.125
  • DescriptionPinchcock Clamp, Heavy-Duty, Nickel-Plated Zinc, 0 to 0.43\" Dia Tubing
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Pinchcocks are designed to quickly start and stop flow and provide complete closure without damaging the tube. Simple squeeze mechanism provides one-handed flow regulation.