Cole-Parmer Pipette Filter Tips 0.5 to 10 µl; PP, clear, graduated, sterile, with filters, 10 racks, 960/pk
Representative image only.
Cole-Parmer Item # EW-25712-73
Cleanroom produced for ready-to-use applications
  • Slim, thin-walled tips feature graduations
  • Filter protects samples and pipettor against contamination
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)10
  • Min Volume (µL)0.5
  • Overall Length (in)1.8125
  • Overall Length (mm)46
  • Body MaterialPP
  • Color CodeClear
  • Filter TypeNon-self sealing
  • Package Of10 racks of 96 each
  • DescriptionPipette Filter Tips 0.5 to 10 µl; PP, clear, graduated, sterile, with filters, 10 racks, 960/pk
Key Features
  • Slim, thin-walled tips feature graduations
  • Filter protects samples and pipettor against contamination
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
More About this Item
Constructed of pure, high-quality polypropylene (PP) without additives, these tips are designed to give high levels of accuracy, precision, and reproducibility. The smooth surface minimizes binding of enzymes and nucleic acids to the tip wall.
The non-self-sealing filter plug prevents aerosols and liquids from being aspirated into the body of the pipettor. Filter is made of pure polyethylene (PE), free from chemical additives, and will not swell.
Tips are available in a storage box with 96-tip rack insert and hinged lid. Order environmentally friendly refill racks separately.