This item is now discontinued. We recommend the following as suggested replacements: EW-24502-68
Cole-Parmer Pipette Tips, 101-1000 ul; 10 racks of 100 each
Representative image only.
Cole-Parmer Item # EW-25711-67
Universal fit pipettor tips for educational and general laboratory applications
  • Precision molded from virgin, chemically-resistant and contamination-free, polypropylene (PP)
  • Tested for fit and function with the most widely used pipettes on the market
  • Fully autoclavable in steam at 121°C for 15 minutes.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)1000
  • Min Volume (µL)101
  • Min Volume (mL)0.101
  • Max Volume (mL)1
  • Package Of10 racks; 1000/pk
  • DescriptionPipette Tips, 101 to 1000 L; 10 racks of 100 each
  • Warranty1 year
1 Year Warranty
Key Features
  • Precision molded from virgin, chemically-resistant and contamination-free, polypropylene (PP)
  • Tested for fit and function with the most widely used pipettes on the market
  • Fully autoclavable in steam at 121°C for 15 minutes.