This item is now discontinued. We recommend the following as suggested replacements: EW-24502-61
Cole-Parmer Pipette Tips 50 to 1000 µl; PP, blue, graduated, bulk bag 1000/pk
Representative image only.
Cole-Parmer Item # EW-25712-33
Cleanroom produced for ready-to-use applications
  • Constructed of pure, high-quality polypropylene without additives
  • Slim, thin-walled tips feature graduations
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)1000
  • Min Volume (µL)50
  • Overall Length (in)2.75
  • Overall Length (mm)70
  • Body MaterialPolypropylene
  • Package OfBag of 1000
  • DescriptionPipette Tips 50 to 1000 µl; PP, clear, graduated, bulk bag 1000/pk
Key Features
  • Constructed of pure, high-quality polypropylene without additives
  • Slim, thin-walled tips feature graduations
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
More About this Item
Designed to give high levels of accuracy, precision, and reproducibility, these polypropylene (PP) tips are suitable for use with most pipettors. The smooth surface minimizes binding of enzymes and nucleic acids to the walls.
Tips are available packaged in a bulk bag or in a storage box with 96-tip rack insert and hinged lid. Order environmentally friendly refill racks separately.