This item is now discontinued. We recommend the following as suggested replacements: EW-10124-23
Cole-Parmer Polystat 13L Heated Circulating Bath with Programmable Controller, 230V
Representative image only.
Cole-Parmer Polystat Item # EW-12121-55
High performance at an affordable price!
  • Store 2 programs with up to 10 steps each — ideal for applications with frequently changing heat loads
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description13L Heated Circulating Bath with Programmable Controller, 230V
  • Warranty2 Years
Two year warrantyISO9001:2000 Certified Supplier
Key Features
  • Store 2 programs with up to 10 steps each — ideal for applications with frequently changing heat loads
  • 2 year warranty
More About this Item
These baths are ideal for industrial or laboratory applications with a wide variety of temperature control requirements. Available in 6-, 13-, and 28-liter tank capacities the compact footprint maximizes bench space. Industrial grade circulating pumps deliver extra pressure providing more efficient heat exchange, better temperature uniformity, and improved reservoir agitation. Added pump pressure provides greater flexibility on where you place the circulator. Long-life high wattage heaters deliver fast fluid heat up for applications requiring rapid fluctuations. Over-temperature and low-level cutouts protect baths from adverse events. Redundant, user-settable over-temperature thermostat provides an added level of security.
Programmable Controller Baths feature the same control and performance characteristics as the Advanced Digital model while offering multistep programming via a twelve-button keypad. Two-line LCD, with contrast control, shows you set point and actual fluid temperature or external temperature and set point. RS-232 interface lets you connect to a PC for datalogging or remote control.