This item is now discontinued. We recommend the following as suggested replacements: EW-12125-65
Cole-Parmer Polystat 28-liter refrigerated bath, 240 VAC Sku1211116
Representative image only.
Cole-Parmer Polystat Item # EW-12111-16
Powerful refrigeration system provides excellent heat load removal across range of operating temperatures
  • The 28.4-liter reservoir is perfect for large-volume applications
  • PID temperature control reduces energy consumption
SPECS & DESCRIPTION
Specifications & Description
  • Description28-liter refrigerated bath, 240 VAC
Includes
lid, two 1/4" NPT(F) x 3/8" hose barb fittings (except models 12108-00 and -05 which come with nylon male adapters for 3/8", 1/4", and 3/16" tubing), and 6-ft power cord with three-prong plug (230 VAC models have European plug).
Key Features
  • The 28.4-liter reservoir is perfect for large-volume applications
  • PID temperature control reduces energy consumption
More About this Item
These baths are ideal for industrial or laboratory applications with a wide variety of temperature control requirements. Available in 6-, 13-, and 28-liter tank capacities the compact footprint maximizes bench space. Robust refrigeration system with metering control provides fast heat removal and stable temperatures. Industrial-grade circulating pumps deliver extra pressure providing more efficient heat exchange, better temperature uniformity, and improved reservoir agitation. Added pump pressure provides greater flexibility on where you place the circulator. Long-life high-wattage heaters deliver fast fluid heat up for applications requiring rapid fluctuations. Overtemperature and low-level cutouts protect baths from adverse events. Secondary, user-settable overtemperature thermostat provides an added level of security. Outlet connections are 1⁄4'' NPT(M).
This bath is perfect for large-volume applications that require precise temperature control. Digital model features RS-232 communication and powerful duplex pump. Features a high-temperature cutoff (adjustable from 60 to 220°C). Inlet and outlet ports on all models are 1/4'' NPT(F) and accept 13-mm ID tubing.