Cole-Parmer Potassium Chloride Electrode Fill Solution w/ AgCl, 4 Molar, 500 mL

Representative image only.
Cole-Parmer Item # EW-88400-59
Get the most out of water analysis equipment
  • Replace or upgrade electrodes, probes, cells, and other accessories

PRODUCT OPTIONS

$60.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassStandards
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7447-40-7, 7761-88-8, 7732-18-5
  • Shelf Life (months)24
  • DescriptionPotassium Chloride Electrode Fill Solution w/ AgCl, 4 Molar, 500 mL

Key Features

  • Replace or upgrade electrodes, probes, cells, and other accessories

More About this Item

Water analysis accessories for Cole-Parmer® meters. Many of these items are compatible with other brands of meter.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat