Cole-Parmer Potassium Iodide, ACS Grade, 500 g

Representative image only.
Cole-Parmer Item # EW-88404-24

$330.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaKI
  • Linear FormulaKI
  • ClassReagent Salts
  • ChemistryAnalytical Reagents;Solutions
  • FormSolid
  • Weight (g)500
  • Formula Weight166.00
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7681-11-0
  • Shelf Life (days)60
  • DescriptionPotassium Iodide, ACS, (500 g)

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat