Cole-Parmer Essentials Precision Stainless Steel Tweezers, Serrated Handle, Strong Tips, AC-SA-B, 110 mm
Representative image only.
Cole-Parmer Essentials Item # EW-07387-14
Ideal for precision handling applications
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
  • Limited Lifetime Warranty
PRODUCT OPTIONS
$13.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 1/4
  • Overall Length (cm)11
  • MaterialStainless Steel Antimagnetic
  • SterileNo
  • StyleAC Sharp, Fine Tips
  • DescriptionPrecision Stainless Steel Tweezers, Serrated Handle, Strong Tips, AC-SA-B, 110 mm
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Antimagnetic and antiacid stainless steel construction
  • Rugged and corrosion-resistant design
  • Limited Lifetime Warranty
More About this Item
These precision stainless steel tweezers offer good corrosion resistance to most chemicals, salts, and acids. Great for lab work such as handling samples or specimens, and observing them under a microscope. The antimagnetic nature of the material makes these tweezers useful in the electronic, watch, and jewelry industries.