Cole-Parmer Premium Grade Iris Tissue Forceps, Straight, 4"

Representative image only.
Cole-Parmer Item # EW-10823-02
Meets the most demanding needs.

PRODUCT OPTIONS

$52.55USD / Each
Often bought with…
Cole-Parmer Liquid Hand Sanitizer, 80% Alcohol; 1 Gallon
$44.00USD / Gallon of 1
$55.00USD / Gallon of 1

SPECS & DESCRIPTION

Specifications & Description

  • Length (in)4
  • Length (cm)10.16
  • DescriptionPremium Grade Iris Tissue Forceps, Straight, 4"

More About this Item

Premium-grade instruments feature the highest quality stainless steel, crafted to the most exacting standards. Autoclavable

Warning — These products are not approved or intended for, and should not be used for, medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat