Cole-Parmer Removable Double-Junction ORP Electrode, Tinned ends
Representative image only.
Cole-Parmer Item # EW-27009-38
Easily remove electrodes in seconds without removing the holder
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
PRODUCT OPTIONS
$306.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • ConnectorTinned ends
  • DescriptionRemovable Double-Junction ORP Electrode, Tinned ends
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for in-line or submersion monitoring
  • 1 year warranty
More About this Item
Use for oxidation/reduction measurements in plating baths, waste-water monitoring, and other general-purpose applications. Platinum band ORP withstands temperatures from 23 to 212ºF (-5 to 100ºC).