Cole-Parmer Replacement motor for drive system 75211-20
Representative image only.
Cole-Parmer Item # EW-75211-22
Permanent-magnet, direct-current (PMDC) motor is built for long-term applications
  • “Soft-start” feature allows gradual, one-second acceleration for smooth flow rate change
  • Continuous-duty drives feature ±2% line/load regulation
  • Controller features IP53-rated housing and single-turn potentiometer for speed control
$589.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • “Soft-start” feature allows gradual, one-second acceleration for smooth flow rate change
  • Continuous-duty drives feature ±2% line/load regulation
  • Controller features IP53-rated housing and single-turn potentiometer for speed control
More About this Item
IP21-rated motor features a welded base for mounting or for free-standing use. Compatible with all Micropump® A-mount pump heads. What’s included: a 6-ft cable for controller-to-motor connection and a 6-ft modular line power cord; the 115 VAC model has a U.S. standard plug and the 230 VAC model has an IEC 320/CEE 22 connector (specify country to obtain correct plug).