Cole-Parmer Replacement Rotor for Personal Microcentrifuge, strip tube
Representative image only.
Cole-Parmer Item # EW-17310-45
Compatible for use with the Cole-Parmer Personal Microcentrifuges
  • Rotors can quickly and easily be interchanged
PRODUCT OPTIONS
$49.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)6000
  • Min Speed (rpm)0
  • Rotor TypeFixed-angle
  • Tube Sizes AcceptedStrip tube
  • Number Of Tubes Accepted2
  • DescriptionReplacement Rotor for Personal Microcentrifuge, strip tube
Key Features
  • Rotors can quickly and easily be interchanged
More About this Item
These replacement rotors are compatible with the 17310-XX series Cole-Parmer personal centrifuges. The model 17310-40 rotor will hold 6 x 1.5 to 2.0 mL tubes. Model 17310-45 rotor will hold 0.4 mL strip PCR tubes. Centrifuges are sold separately.