Cole-Parmer RS-232 Serial Adapter, DB9 and RJ11
Representative image only.
Cole-Parmer Item # EW-93284-20
Optimize performance of your Cole-Parmer process control system
  • Keep your process control system running properly with these accessories
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRS-232 Serial Adapter, DB9 and RJ11
  • Warranty3 Years
3 Year Warranty
Key Features
  • Keep your process control system running properly with these accessories
  • 3 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Cole-Parmer process control system.
Converts the serial output of Cole-Parmer® flow and panel meters to an unbalanced, full-duplex RS-232 signal. Since it is an external device, one adapter in conjunction with the free MasterView® software, can be used to program and calibrate an infinite number of meters.