Cole-Parmer Essentials Safety Goggles; 12 box/case
Representative image only.
Cole-Parmer Essentials Item # EW-16392-24
For protection against chemical splash and other contaminants
  • Economical and comfortable goggle
  • 99.99% UV protection (UV-A, UV-B, UV-C)
  • Fits over most prescription glasses
  • Polycarbonate antiscratch lens with antifog coating
  • Ultrasonic-sealed indirect HDPE vent caps
  • 13mm Headband for user comfort
  • Class 1 high optical quality lenses
  • Dust proof, wind proof and splash protection
  • 3 year warranty
PRODUCT OPTIONS
$536.50USD / Case of 120
BUY MORE & SAVE
  • 1-2$536.50USD / Case of 120
  • 3+$482.85USD / Case of 120
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorClear
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceEN166:2001; ANSI Z87.1:2015
  • DescriptionSafety Goggles, 10 pcs/box; 12 box/case
  • Warranty3 year
3 Year Warranty
Key Features
  • Economical and comfortable goggle
  • 99.99% UV protection (UV-A, UV-B, UV-C)
  • Fits over most prescription glasses
  • Polycarbonate antiscratch lens with antifog coating
  • Ultrasonic-sealed indirect HDPE vent caps
  • 13mm Headband for user comfort
  • Class 1 high optical quality lenses
  • Dust proof, wind proof and splash protection
  • 3 year warranty
More About this Item
Certifications:
• CE certified
• EN166:2001 compliant
• ANSI Z87.1-2015 compliant