Cole-Parmer Saw-Tooth Generator, 195mm x 20mm, Course, Wide

Representative image only.
Cole-Parmer Item # EW-04727-43
Choose the generator based on the dispersion, tissue type, and or sample size
  • Autoclavable, stainless steel construction
  • Easily disassembled for cleaning
  • 20 mm diameter x 195 mm length
  • Wide window for large partical disruption

PRODUCT OPTIONS

$1,900.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Dispersion TypeCourse
  • Use with Generators04727-28 and 04727-29
  • Min Sample Size (mL)100
  • Max Sample Size (mL)2000
  • Tip Diameter (mm)20
  • DescriptionSaw-Tooth Generator, 195mm x 20mm, Course, wide

Key Features

  • Autoclavable, stainless steel construction
  • Easily disassembled for cleaning
  • 20 mm diameter x 195 mm length
  • Wide window for large partical disruption

More About this Item

These generator probes come with stainless steel upper bearings. Applications include large volume emulsions, suspensions, and dispersions for liquid/liquid or solid/liquid.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat