Cole-Parmer Scope/Laser Sight Infrared Thermometer; Class II, Single, 105: 1
Representative image only.
Cole-Parmer Item # EW-39800-35
Feature adjustable emissivity, recall, HI/LO alarms, MIN, MAX, DIF, AVG, and data output Take temperature measurements in both bright-light and low-light areas with new model 39800-35
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)105:1
  • EmissivityAdjustable
  • Sighting TypeScope and single-point laser
  • Laser ClassClass II
  • Min Temperature (° F)-20
  • Max Temperature (° F)2200
  • Min Temperature (° C)-30
  • Max Temperature (° C)1200
  • Accuracy±1% of reading or ±1.5°F (±1.0°C), whichever is greater
  • Resolution0.1°; 1° above 1000°
  • Response time<700 milliseconds
  • RS-232 InterfaceYes
  • Output1mV/°F or 1mV/°C
  • Display Type0.38\", 4-digit LCD
  • BatteryAA Battery (4)
  • DescriptionScope/Laser Sight Infrared Thermometer; Class II, Single, 105: 1
  • Warranty1 year
1 Year WarrantyLaser Class II
Key Features
  • 1 year warranty
More About this Item
These infrared thermometers offer a variety of aiming systems to fit your exact requirements. Scope models feature a through-the-lens optical system and a circular reticle to precisely define the target area.
Use a laser sighting model to determine the center of the area you are measuring. Laser spots on the target indicate the approximate diameter. Use single-laser models for longrange measurements. Dual and crossed laser models have lasers that are aligned with the thermometer's optics. New model 39800-35 offers both scope and laser sighting.
Choose from two laser types-class IIIa laser models for maximum brightness, or class II models where regulations require use of lowpower lasers.
Lock the trigger for long-term monitoring. Store up to 100 temperature readings at selectable time intervals with built-in clatalogger. Configure output as either RS-232 or as a one millivolt per degree analog output (cables and software sold separately below).
Thermometers display minimum, maximum, differential temperature, and average temperature in °F or °C. Other features include adjustable emissivity, recall, backlit display, and an audible and visual alarm. Each thermometer includes four AA batteries, padded nylon carrying case, shoulder strap, and instructions. Adapters for 115 or 230 VAC operation are available—order below. Shpg wt 5.0 lb (2.3 kg).