Cole-Parmer SD7500CD pH Sensor, Differential, Self-Powered Preamp
Representative image only.
Cole-Parmer Item # EW-27400-69
Longer working lifetime with less maintenance
  • Corrosion resistant through buffered reference cell solution
  • Quick and easy maintenance with replacement salt bridge
  • Improved chemical compatibility with PPS electrode body
  • 2 year warranty
PRODUCT OPTIONS
$937.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSalt bridge
  • Electrode Body MaterialPPS
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)85
  • Temperature CompensationYes
  • Max Pressure (PSI)100
  • Thread Size NPT(M) (in)1 1/2
  • ATC Connector5 conductor plus shield
  • ATC ElementPT1000 RTD
  • Connector5 conductor plus shield
  • Cable Length (m)6.1
  • DescriptionpH Sensor, Differential, Self-Powered Preamp
  • Warranty2 year
Two year warranty
Key Features
  • Corrosion resistant through buffered reference cell solution
  • Quick and easy maintenance with replacement salt bridge
  • Improved chemical compatibility with PPS electrode body
  • 2 year warranty
More About this Item
The differential technology of these sensors provides advanced accuracy by virtually eliminating ground loops. In addition, the automatic temperature compensation of the sensors allows for precise measurement in temperature changing environments.
These sensors are made with a PPS body and salt bridge, PVDF and ceramic junction, Viton® O-rings, and a titanium solution ground. The pH sensors have a protected glass bulb while the ORP sensors have a platinum band. The durable construction allows for enhanced chemical compatibility.
These differential pH sensors have two reference electrodes for differential measurement of either pH or ORP with respect to a third metal ground electrode. These feature a reference cell which is a measuring electrode in a buffered pH 7.0 buffer solution. Using the buffered reference cell rather than the traditional silver/silver chloride reference resists contamination by the sample, improves accuracy, and extends the sensor life. Sensor life is also extended with the replacement salt bridge feature and refill solution, which avoids having to replace the entire sensor.