Cole-Parmer SDA-7010 Salt bridge for SD7000CD & -ORP, 10 each
Representative image only.
Cole-Parmer Item # EW-27400-73
Accessories for the Cole-Parmer pH and ORP differential sensors
  • Quick and easy maintenance with optional replacement salt bridge
PRODUCT OPTIONS
$409.25USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeReplacement Salt Bridge
  • MaterialPPS
  • DescriptionSalt bridge for SD7000CD & -ORP, 10 each
Key Features
  • Quick and easy maintenance with optional replacement salt bridge
More About this Item
Sensor life is extended with the replacement salt bridge feature and refill solution, which avoids having to replace the entire sensor. Choose from the replacement salt bridge in packs of either 1, 3, 10 10 each. The standard cell refill solution is available in three sizes as well.