Oakton by Cole-Parmer® sealed pH Electrode, Double-Junction for Orion A Series Meter
Representative image only.
Oakton Item # EW-27508-22
Works with meters you already have
  • Combine the pH reference, pH half-cell, and temperature sensor in one 12-mm diameter electrode body
  • 1 year warranty
PRODUCT OPTIONS
$177.30USD / Each
$197.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSealed
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • ConnectorBNC
  • DescriptionSealed pH Electrode, Double-Junction for Orion A Series Meter
  • Warranty1 year
1 Year Warranty
Key Features
  • Combine the pH reference, pH half-cell, and temperature sensor in one 12-mm diameter electrode body
  • 1 year warranty
More About this Item
Epoxy-body pH electrodes compatible with Orion A-Series meters.