Oakton by Cole-Parmer® Self-Cleaning Flat Surface ORP Probe, DJ/Ryton/Pt/10'/BNC
Representative image only.
Oakton Item # EW-27003-50
Flat electrode surface prevents buildup of solids
  • Flat surface prevents buildups, while fluid flow cleans surface for longer life
  • Use with most meters and controllers—BNC connector and ATC element work with a variety of units
  • Double junction for difficult applications
  • Choose either CPVC housing or chemical-resistant Ryton® PPS housing
  • 1 year warranty
PRODUCT OPTIONS
$276.00USD / Each
BUY MORE & SAVE
  • 1-5$276.00USD / Each
  • 6-11$248.40USD / Each
  • 12-23$234.60USD / Each
  • 24+$220.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSelf-cleaning
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Temperature CompensationYes
  • ConnectorBNC
  • DescriptionSelf-Cleaning Flat Surface ORP Probe, DJ/Ryton/Pt/10'/BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Flat surface prevents buildups, while fluid flow cleans surface for longer life
  • Use with most meters and controllers—BNC connector and ATC element work with a variety of units
  • Double junction for difficult applications
  • Choose either CPVC housing or chemical-resistant Ryton® PPS housing
  • 1 year warranty
More About this Item
Use these unique double-junction electrodes in your most difficult ORP applications. Flat surface does not collect suspended solids--fluid flow across the electrode provides a cleaning action that extends electrode life and improves performance. Annular junction reduces reference cell fouling. Choose either a CPVC housing (max temp 80°>C) or chemical resistant Ryton® housing (max temp 100°>C). Both types have 10-ft cables and can be used at pressures up to 100 psi. BNC connectors match a variety of meters and controllers. Install electrodes in tee fittings--3/4'' NPT(M) thread and 2'' insertion depth.