Oakton by Cole-Parmer® self-cleaning pH electrode, Ryton housing, 10-ft cable, BNC
Representative image only.
Oakton Item # EW-27003-20
No more solid buildups
  • Flat surface prevents buildups, while fluid flow cleans surface for longer life
  • Use with most meters and controllers—BNC connector and ATC element work with a variety of units
  • Double junction for difficult applications
  • 1 year warranty
PRODUCT OPTIONS
$231.00USD / Each
BUY MORE & SAVE
  • 1-5$231.00USD / Each
  • 6-11$207.90USD / Each
  • 12-23$196.35USD / Each
  • 24+$184.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Min pH (pH)12
  • Max Temperature (° C)100
  • Max Pressure (PSI)100
  • Thread Size NPT(M) (in)3/4
  • ATC ElementNone
  • ConnectorBNC
  • Cable Length (m)3
  • Descriptionself-cleaning pH electrode, Ryton housing, 10-ft cable, BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Flat surface prevents buildups, while fluid flow cleans surface for longer life
  • Use with most meters and controllers—BNC connector and ATC element work with a variety of units
  • Double junction for difficult applications
  • 1 year warranty
More About this Item
Choose either CPVC housing or chemical-resistant Ryton® PPS housing