Cole-Parmer Semi-domed pH electrode, PPS housing, 100-Ohm Pt RTD, 10-ft cable, BNC connector
Representative image only.
Cole-Parmer Item # EW-27021-02
Won’t fail even in tough industrial environments
  • Unique design protects bulb
  • Reduce electrolyte leakage and sample contamination—stand up to viscous samples better than PTFE junctions
  • Extended life in severe applications
  • Graphite-tipped models eliminate electrical noise from your system
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Min pH (pH)0
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)110
  • Temperature CompensationYes
  • Max Pressure (PSI)150
  • ATC ConnectorMini-audio plug
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • Length (mm)150
  • Length (in)5 29/32
  • DescriptionSemi-domed pH electrode, PPS housing, 100-Ohm Pt RTD, 10-ft cable, BNC connector
  • Warranty1 year
1 Year Warranty
Key Features
  • Unique design protects bulb
  • Reduce electrolyte leakage and sample contamination—stand up to viscous samples better than PTFE junctions
  • Extended life in severe applications
  • Graphite-tipped models eliminate electrical noise from your system
  • 1 year warranty
More About this Item
Choose from a variety of unique electrode designs for use in tough industrial environments. All models feature BNC pH connectors to match a variety of meters and controllers. Electrodes with temperature compensation feature 100-Ω RTD temperature elements with mini-audio plug connections
These double-junction electrodes have a revolutionary annular ceramic reference junction and a uniform pore size that reduces contamination and clogging.