This item is now discontinued. Please browse similar items
Cole-Parmer Shop Microscope; 40x magnification
Representative image only.
Cole-Parmer Item # EW-03890-20
Take anywhere on the job - ideal for lab, field, or shop work
SPECS & DESCRIPTION
Specifications & Description
  • Magnification40x
  • Maximum Magnification40x
  • Head ConfigurationMonocular
  • Working Distance23 mm
  • Field of View4.5 mm
  • DescriptionShop Microscope; 40x magnification
Includes
plastic carrying case, dust cover, and two AA batteries
More About this Item
Small size and rugged carrying case make it easy to take these shop microscopes anywhere! The cast aluminum frame with gray enamel finish measures only 9''H. Wide-field 10x eyepiece features a direct-reading reticle with both inch and metric scales. Simply turn the eyepiece to bring the reticle into sharp focus - no calibration is needed. Dual knobs with rack and pinion focusing let you quickly refine the image. Penlight illuminator is fully adjustable in all directions by means of a ball joint; remove it for use elsewhere.