Cole-Parmer Side-Mount Tuff-Tip pH Probe, SJ/PVDF/1kOhm RTD; Spade/BNC
Representative image only.
Cole-Parmer Item # EW-27011-44
Durable Tuff-Tip with ability to mount sideways
  • No more breakage—tip design protects bulb
  • Full range; measure up to pH of 14
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialPVDF
  • Min pH (pH)0
  • Max pH (pH)13
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • Thread Size NPT(M) (in)1/2
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionSide-Mount Tuff-Tip pH Probe, SJ/PVDF/1kOhm RTD; Spade/BNC
  • Warranty1 year
1 Year WarrantyKeep a Spare
Key Features
  • No more breakage—tip design protects bulb
  • Full range; measure up to pH of 14
  • 1 year warranty
More About this Item
The tip design protects the bulb from breakage and the gel internal fill solution allows electrode to be installed where others cannot. Rugged PVDF housing with 1/2'' NPT (M) threads withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Use for measurements up to pH 13. Models include a 10-ft cable with BNC pH connector. Insertion depth 1''.