Cole-Parmer Essentials Sieve; No 100, Half-Height, 12", Brass/Brass, NIST-Traceable Calibration Certificate
Representative image only.
Cole-Parmer Item # EW-59991-66
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass frame, brass wire
  • U.S. Standard Screen Size (No.)100
  • Tyler Standard Screen Size (mesh)100
  • Sieve Opening (mm)0.149
  • Outside Diameter (in)12
  • Height (InchShort)1.625
  • DescriptionSieve; No 100, Half-Height, 12\", Brass/Brass, NIST Cal Cert
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Sieve is serialized, comes with a test sieve certificate, and meets ASTM E11 specifications. Includes an NIST-traceable calibration report supplied by the manufacturer.