Cole-Parmer Essentials Sieve; No 12, Half-Height, 8", Brass/Brass, NIST-Traceable Calibration Certificate
Representative image only.
Cole-Parmer Essentials Item # EW-59981-52
Sieve openings are measured with a computerized video imaging system
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass frame, brass wire
  • U.S. Standard Screen Size (No.)12
  • Tyler Standard Screen Size (mesh)10
  • Sieve Opening (mm)1.68
  • Outside Diameter (in)8
  • Height (InchShort)1
  • DescriptionSieve; No 12, Half-Height, 8\", Brass/Brass, NIST Cal Cert
  • Warranty1 Year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • 1 year warranty
More About this Item
Sieve is serialized, comes with a test sieve certificate, and meets ASTM E11 specifications. Includes an NIST-traceable calibration report supplied by the manufacturer.