Cole-Parmer Essentials Sieve; No 140, Full-Height, 12", SS/SS, NIST-Traceable Calibration Certificate
Representative image only.
Cole-Parmer Essentials Item # EW-59994-68
  • 1 year warranty
PRODUCT OPTIONS
$397.50USD / Each
$157.00 - $230.00USD / Each
for an additional
$157.00USD / Each
for an additional
$230.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)140
  • Tyler Standard Screen Size (mesh)150
  • Sieve Opening (mm)0.105
  • Outside Diameter (in)12
  • Height (InchShort)3.25
  • DescriptionSieve; No 140, Full-Height, 12\", SS/SS, NIST Cal Cert
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Sieve is serialized, comes with a test sieve certificate, and meets ASTM E11 specifications. Includes an NIST-traceable calibration report supplied by the manufacturer.