Dow Corning Silicone Rubber Bonding Cement, –85 to 450°F; 3 oz Tube
Representative image only.
Dow Corning Mfr # RTV-732-WHITE – Item # EW-03125-80
Silicone rubber bonding cement for All-Purpose Silicone Rubber Blanket Heaters
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSilicone rubber bonding cement, 3 oz tube, –85 to 450°F
More About this Item
Cures in 15 to 20 minutes at room temperature and has a temperature range of –85 to 450°F (–65 to 232°C).