Cole-Parmer Silicone Sanitary Gasket, 3/4" Tri-Clamp; 10/Pack
Representative image only.
Cole-Parmer Item # EW-30548-00
$20.30USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPlatinum-cured silicone
  • Connection TypeSanitary
  • DescriptionSilicone Sanitary Gasket, 3/4" Tri-Clamp; 10/Pack
Certificate of Compliance
More About this Item
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match your specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class IV and cytotoxicity criteria. Contact Cole-Parmer for other sizes or materials.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from -58 to 446°F (-50 to 230°C).
Viton offers excellent chemical resistance, especially to many acids and alkalies.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
Cole-Parmer Silicone Sanitary Gasket, 3/4" Tri-Clamp; 10/Pack
4.0
(based on 1 review)
Reviewed by 1 customer
Displaying review 1
Back to top
4.0
Great Product
By 
from Ottawa On
Pros
  • Multi use
  • Sealed well
Cons
    Best Uses
    • Filtering
    Comments about Cole-Parmer Silicone Sanitary Gasket, 3/4" Tri-Clamp; 10/Pack:
    I like the packaging size
    • Was this a gift?:
    • No
    Displaying review 1
    Back to top
    Q&A
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    5 Questions | 5 Answers
    Displaying questions 1-5
    • Q:
      Are these gamma stable and to what specification?
      Do you have a working pressure pre and post gamma?
      Asked on 1/19/2016 by Anonymous
      1 answer
      • STAFF EXPERT
        A:
        These odorless, tasteless, non-toxic hygienic seals maintain product integrity and can be autoclaved, irradiated and gas sterilized. For SIP procedures, this gasket is suitable up to 30 psi at 123C.
        Answered on 1/22/2016 by Danielle from Technical Support
    • Q:
      can this be certified as TSE free?
      Asked on 6/23/2014 by Phil from Oxford
      1 answer
      • STAFF EXPERT
        A:
        This item is TSE/BSE animal origin free. We can also supply a statement for this if needed once ordered.
        Answered on 6/27/2014 by Jose from Technical Support
    • Q:
      EW-30548-00
      Sanitary Gasket, Silicone, 3/4", 10/pk
      can this product be autoclaved at 121°C?
      Asked on 3/8/2011 by Anonymous
      1 answer
      • Staff Reviewer
        A:
        Yes, these are autoclavable. 121C is a standard autoclave temperature.
        Answered on 3/9/2011 by Jen from Technical Support
    • Q:
      Are these measured using the inside diameter or the outside?
      Asked on 2/21/2011 by mlfmaine from Winslow, Maine
      1 answer
      • STAFF EXPERT
        A:
        The gasket is actually made to fit a 3/4" sanitary connection. The ID (bore) of a 3/4" sanitary fitting is 5/8" and the OD of the flange is just under 1", or 63/64".
        Answered on 2/21/2011 by Jack from Technical Support
    • Q:
      Can clear silicone tri-clamp gaskets (.75in) be sterilized by autoclaving or ETOH treatment
      Asked on 5/6/2010 by Bill from Cincy OH
      1 answer
      • STAFF EXPERT
        A:
        Silicone can be sterilized by ETO or autoclave.
        Answered on 5/10/2010 by David from Tech Support
    Displaying questions 1-5
    Do you have a question about this product? 