Cole-Parmer Size 1 high-flow polypropylene filter bag; 200°F (93°C) maximum
Representative image only.
Cole-Parmer Item # EW-01523-46
Wide range of options for temperature, flow rate, and filtration quality
  • Constructed of Polypropylene Felt
$5.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolyester
  • Micron Rating25
  • Flow Rate90 GPM
  • Filter Length (in)16.5
  • Filter Diameter (in)7
  • Filtration Area (ft²)2
  • Filtration Area (in²)288
  • Filtration Area (m²)0.186
  • DescriptionSize 1 high-flow polypropylene filter bag; 200°F (93°C) maximum
Key Features
  • Constructed of Polypropylene Felt
More About this Item
Designed for higher flow with an average efficiency at the rated pore size. Polypropylene felt is ideal for general-purpose filtration. The bags are glazed to minimize fiber migration. Polypropylene felt filter bags are rated for 200°F (93°C) maximum.