Cole-Parmer Size 3 high-flow nylon monofilament filter bag; 300°F (149°C) maximum
Representative image only.
Cole-Parmer Item # EW-01524-58
Wide range of options for temperature, flow rate, and filtration quality
  • Constructed of nylon
$7.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaNylon
  • Micron Rating300
  • Flow Rate30 GPM
  • Filter Length (in)8
  • Filter Diameter (in)4.125
  • Filtration Area (ft²)0.5
  • Filtration Area (in²)72
  • Filtration Area (m²)0.046
  • DescriptionSize 3 high-flow nylon monofilament filter bag; 300°F (149°C) maximum
Key Features
  • Constructed of nylon
More About this Item
Designed for higher flow with an average efficiency at the rated pore size. The bags are glazed to minimize fiber migration. Nylon monofilament is a highstrength bag for handling a relatively more aggressive, cruder filtration application. These have a temperature rating of 300°F (149°C) maximum.