Cole-Parmer Size 4 high-flow polyester filter bag; 300F (149C) max, 25 um porosity
Representative image only.
Cole-Parmer Item # EW-01524-84
Wide range of options for temperature, flow rate, and filtration quality
  • Constructed of Polyester Felt
PRODUCT OPTIONS
$5.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolyester
  • Micron Rating25
  • Flow Rate40 GPM
  • Filter Length (in)14
  • Filter Diameter (in)4.125
  • Filtration Area (ft²)1
  • Filtration Area (in²)144
  • Filtration Area (m²)0.093
  • DescriptionSize 4 high-flow polyester filter bag; 300F (149C) max, 25 um porosity
Key Features
  • Constructed of Polyester Felt
More About this Item
Designed for higher flow with an average efficiency at the rated pore size. Polyester felt is ideal for general-purpose filtration. The bags are glazed to minimize fiber migration. Choose polyester for higher temperature applications, rated for a temperature range of 300°F (149°C) maximum.