Cole-Parmer Slow Emptying Device, for 1.6" OD PE and PVC Bailers
Representative image only.
Cole-Parmer Item # EW-99157-06
Enhance the versitility of your bailers with these accessories
  • Replacement dipper shaft
  • Slow emptying devices
PRODUCT OPTIONS
$75.00USD / Each
BUY MORE & SAVE
  • 1-4$75.00USD / Each
  • 5-9$67.50USD / Each
  • 10+$63.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithBailers 99157-00 to -05, -40, -42, -44
  • DescriptionSlow Emptying Device, for 1.6\" OD PE and PVC Bailers
Key Features
  • Replacement dipper shaft
  • Slow emptying devices
More About this Item
Model 06282-19 replacement dipper shaft is compatible with Cole-Parmer dipper series 06282-11 to -17. Slow Emptying Devices reduce flow from bailer, minimizing agitation for VOC analysis. Model 99157-06 device is compatible with bailers 99157-00, -01, -02, -03, -04, -05, -40, -42, and -44. Model 99157-16 device is compatible with bailers 99157-12 and -13.